Learn More
3M™ Petrifilm™ High-Sensitivity Coliform Count Plate
Accommodates 5 mL samples and detection of low levels of confirmed coliforms in 24 hours.
Brand:  3M™ Petrifilm™ 7100039357
Additional Details : CAS Number : 9000-30-0 Weight : 0.01200kg
 View more versions of this product
Product Code. 17223754
  • 133.56 € / Pack of 50
Estimated Shipment: 13-08-2024
to see stock.
Add to basket
Description
Description
  • Coliform results in 24 hours.
  • Plates are sample-ready, eliminating all the time-consuming steps of preparing media/agar dishes.
  • Unique plate design to trap the gas produce by coliform
  • This plate has been awarded Official Methods of Analysis (OMA) #996.02 for Dairy products and NF VALIDATION certificate: 3M-01/7-03/99 for Human food products by AFNOR Certification.
  • Food processors and contract labs worldwide trust 3M Petrifilm Plates to provide accurate, efficient, consistent results.
Specifications
Specifications
Environmental Test
50