Learn More
Neogen Petrifilm™ High-Sensitivity Coliform Count Plate
Product Code. 17253754
Encompass
Change view
Click to view available options
Quantity:
50
500
Unit Size:
Pack of 50
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17253754 500 Pack of 500
17223754 50 Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17253754 Supplier Neogen Petrifilm™ Supplier No. 7100039398
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Accommodates 5 mL samples and detection of low levels of confirmed coliforms in 24 hours.
  • Coliform results in 24 hours.
  • Plates are sample-ready, eliminating all the time-consuming steps of preparing media/agar dishes.
  • Unique plate design to trap the gas produce by coliform
  • This plate has been awarded Official Methods of Analysis (OMA) #996.02 for Dairy products and NF VALIDATION certificate: Neogen-01/7-03/99 for Human food products by AFNOR Certification.
  • Food processors and contract labs worldwide trust Neogen Petrifilm Plates to provide accurate, efficient, consistent results.
Specifications
Product Type Count Plate
Test Type Food and Beverage, Environmental Test
Quantity 500