Learn More
High Temperature Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified, viscosity 149.2 mPa.s (60 °C), Paragon Scientific™
Description
- Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation
- Kinematic (cSt), dynamic viscosity (mPa·s) and density (g/mL) given at all temperatures
- Standard Pack Size 500mL
- Fully traceable to National Standards
- Ensures full compliance to ASTM & IP test method protocol
- Tamper evident security packaging
- 2 Year shelf life
- Manufactured in the United Kingdom
Specifications
Specifications
| Quantity | 500 mL |
Nominal values quoted. Certified Values will vary from batch to batch.