missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiLoad™ 16/600 Superose™ 6 Prep Grade Preparative SEC Column

Product Code. 16022082 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16022082 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16022082 Supplier Cytiva Supplier No. 29323952
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A prepacked SEC column for high-resolution purification of large biomolecules and protein complexes with sample volumes up to 5 mL

  • Can be used with a standalone pump or a chromatography system
  • Purification is possible across an exceptionally broad molecular-weight range
  • Separations can be scaled up using Superose™ 6 Prep Grade Resin
  • Resin provides good resolution separations of proteins and other biomolecules according to size
  • Fittings (1/16 in.) for direct connection to ÄKTA systems and other chromatography systems
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Ea.
Flow Rate 1.6 mL/min
Bed Volume 120 mL
Product Type Superose Pre-packed Column
Diameter (Metric) Inner 16 mm
For Use With (Application) Size Exclusion Chromatography (SEC)
Height (Metric) 600 mm
Sample Type <5 mL
Includes Dismantling Tool, Net Ring (10 μm), Stop Plugs, O-Ring, Support Screen, and Instructions
Content And Storage Store at 4°C to 30°C
Stationary Phase Superose™ 6 Prep Grade (pg) Resin
Show More Show Less
compliance-icons
Product Identifier
  • HiLoad 16/600 Superose 6 pg
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.