missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HIN-1 Monoclonal Antibody (368203)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA561231

Product Code. 30535896

  • 459.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

HIN-1, also known as High In Normal-1 or Secretoglobin Family 3A Member 1 (SCGB3A1), is a gene located on chromosome 5q33.3. It encodes a protein that is predominantly expressed in normal mammary epithelial cells and is thought to function in epithelial cell signaling, contributing to cellular differentiation and maintaining normal tissue architecture. The HIN-1 gene is often silenced or downregulated in various cancers, particularly breast cancer, due to hypermethylation of its promoter region, suggesting a potential tumor suppressor role. Loss of HIN-1 expression has been implicated in the progression of malignancies by disrupting normal cell signaling pathways, thereby facilitating oncogenesis. Given its significant downregulation in cancerous tissues, HIN-1 is being explored as a biomarker for early cancer detection and may provide insights into novel therapeutic strategies that aim to restore its expression and normal cellular functions.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HIN-1
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q920D7
SCGB3A1
E. coli-derived recombinant mouse HIN-1/SCGB3A1, Phe22-Gly104.
100 μg
Primary
Mouse
Antibody
IgG2a
Western Blot
368203
Unconjugated
SCGB3A1
cytokine high in normal-1; cytokine HIN-1; High in normal 1; HIN1; HIN-1; LU105; pneumo secretory protein 2; PNSP2; PnSP-2; SCGB3A1; Secretoglobin family 3A member 1; secretoglobin, family 3A, member 1; UGRP2; UGRP2MGC87867; UNQ629/PRO1245; Uteroglobin-related protein 2
Rat
Protein A/G
RUO
68662
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.