missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beskrivning
- Long-term chemical and physical stability from hydrophilic, rigid allyl dextran/bisacrylamide matrix
- Recommended linear flowrate 5 to 15cm/hr.
Designed for separating peptides and small proteins
Specifikationer
Specifikationer
| Quantity | Each |
| Flow Rate | <0.5 mL/min. |
| Particle Size | 25 to 75 μm |
| For Use With (Equipment) | Low to medium pressure AKTA™ systems |
| Bed Volume | 120 mL |
| Product Type | Sephacryl HR Column |
| Diameter (Metric) Inner | 1.6cm |
| For Use With (Application) | Separation of biomolecules with maximum recovery and minimum nonspecific adsorption with long term chemical and physical stability resulting from the hydrophilic, rigid Allyl Dextran/Bisacrylamide matrix |
| Height (English) | 23.62 in. |
| Height (Metric) | 60cm |
| Visa mer |
Säkerhet och hantering
Produktbeteckning
- HiPrep 16/60 Sephacryl S-100 HR
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?