missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiPrep™ Sephacryl™ S-300 HR Columns

Product Code. 10702817 Shop All Cytiva Products
Change view
Click to view available options
Flow Rate:
<0.5 mL/min.
<1.3 mL/min.
Bed Volume:
120mL
320mL
Diameter (Metric) Inner:
1.6cm
2.6cm
Format:
Bed Dimensions: 16x60
Bed Dimensions: 26x60
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Bed Volume Diameter (Metric) Inner Format Unit Size
10702817 <1.3 mL/min. 320mL 2.6cm Bed Dimensions: 26x60 Each
10616695 <0.5 mL/min. 120mL 1.6cm Bed Dimensions: 16x60 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10702817 Supplier Cytiva Supplier No. 17119601
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Maximum recovery and minimum nonspecific adsorption

  • Long-term chemical and physical stability from hydrophilic, rigid allyl dextran/bisacrylamide matrix
  • Recommended linear flowrate 5 to 15cm/hr.
  • Designed for purifying antibodies, serum proteins and midsize proteins

TRUSTED_SUSTAINABILITY

Specifications

Quantity Each
Flow Rate <1.3 mL/min.
Particle Size 25 to 75 μm
For Use With (Equipment) Low to medium pressure AKTA™ systems
Bed Volume 320mL
Product Type Sephacryl HR Column
Format Bed Dimensions: 26x60
Diameter (Metric) Inner 2.6cm
For Use With (Application) Separation of biomolecules with maximum recovery and minimum nonspecific adsorption with long term chemical and physical stability resulting from the hydrophilic, rigid Allyl Dextran/Bisacrylamide matrix
Height (English) 23.62 in.
Height (Metric) 60cm
Material Polypropylene
Max. Pressure 22psi
pH Range 3 to 11
Support Type Allyl Dextran and N, N'-methylenebisacrylamide
Sample Type Antibodies, serum proteins and mid-size proteins
Content And Storage 4° to 30°C
Show More Show Less
compliance-icons
Product Identifier
  • HiPrep 26/60 Sephacryl S-300 HR
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.