missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ HIST1H2BK Antibody (SR3153), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27749-50ul
This item is not returnable.
View return policy
Description
HIST1H2BK Monoclonal antibody specifically detects HIST1H2BK in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| HIST1H2BK | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 85236 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| SR3153 | |
| Western Blot 1:5000-1:20000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:30 | |
| H2B histone family, member T, H2B K, H2B/S, H2BFAiii, H2BFT, H2bk, HIRA-interacting protein 1, HIRIP1, histone cluster 1, H2bk, histone family member, histone H2B type 1-K, MGC131989 | |
| A synthesized peptide derived from human HIST1H2BK (Uniprot #: O60814) | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction