missing translation for 'onlineSavingsMsg'
Learn More

Histamine phosphate, MedChemExpress

Product Code. 16452019
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16452019 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16452019 Supplier Medchem Express Supplier No. HYA0129

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Histamine (phosphate) diphosphate is a potent agonist of histamine receptors and vasodilator.

Histamine (phosphate) diphosphate is a potent agonist of histamine receptors and vasodilator.

Specifications

Chemical Name or Material Histamine phosphate
Physical Form Solid
CAS 51-74-1
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C5H15N3O8P2
For Use With (Application) Cancer-programmed cell death
Synonym Histamine diphosphate
Solubility Information H2O : 100 mg/mL (325.58 mM; Need ultrasonic) ∣DMSO : 1.43 mg/mL (4.66 mM; Need ultrasonic)
SMILES NCCC1=CNC=N1.O=P(O)(O)O.O=P(O)(O)O
Molecular Weight (g/mol) 307.14
Formula Weight 307.14
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.