missing translation for 'onlineSavingsMsg'
Learn More

Histone Deacetylase 2/HDAC2 Antibody (9E9), Novus Biologicals™

Rabbit Monoclonal Antibody

343.00 € - 533.00 €

Specifications

Antigen Histone Deacetylase 2/HDAC2
Clone 9E9
Dilution Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30026658
View Documents
Novus Biologicals
NBP3-26238-50ul
50 μL
343.00 €
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30038049
View Documents
Novus Biologicals
NBP3-26238-100ul
100 μL
533.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Histone Deacetylase 2/HDAC2 Monoclonal antibody specifically detects Histone Deacetylase 2/HDAC2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications

Specifications

Histone Deacetylase 2/HDAC2
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Monoclonal
Purified
RUO
Human
EC 3.5.1.98, HD2, histone deacetylase 2, RPD3, transcriptional regulator homolog RPD3, YAF1, YY1-associated factor 1
A synthesized peptide derived from Human Histone Deacetylase 2/HDAC2 [UniProt Q92769]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
9E9
Western Blot, ELISA, Immunohistochemistry
Unconjugated
Rabbit
Breast Cancer, Cancer, Chromatin Research, Epigenetics, Phospho Specific, Stem Cell Markers
PBS, pH 7.4, 150mM NaCl and 50% glycerol
3066
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.