missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone Deacetylase 2/HDAC2 Antibody (OTI7E10), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70881H
This item is not returnable.
View return policy
Description
Histone Deacetylase 2/HDAC2 Monoclonal specifically detects Histone Deacetylase 2/HDAC2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Histone Deacetylase 2/HDAC2 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| HDAC2 | |
| Recombinant protein fragment corresponding to amino acids 415-488 of human Histone Deacetylase 2/HDAC2 (NP_001518) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI7E10 | |
| Western Blot, Immunohistochemistry | |
| EC 3.5.1.98, HD2, histone deacetylase 2, RPD3, transcriptional regulator homolog RPD3, YAF1, YY1-associated factor 1 | |
| Mouse | |
| Affinity Purified | |
| Breast Cancer, Cancer, Chromatin Research, Epigenetics, Histone Deacetylases, Phospho Specific, Stem Cell Markers | |
| 3066.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction