Learn More
Histone H2AX Antibody (CL5157) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21157JF525
Product Code. 30287057
  • 759.00 € / 0.10mL
Description
Histone H2AX Monoclonal antibody specifically detects Histone H2AX in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Histone H2AX
Monoclonal
Janelia Fluor 525
H2A.X, H2A/X, H2AFX
This antibody was developed using a synthetic peptide derived from P16104, with the exact immunogen sequence remaining proprietary.
0.1 mL
Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Mitotic Regulators, Phospho Specific
3014
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL5157
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified