missing translation for 'onlineSavingsMsg'
Learn More

Histone H2AX, p Ser139 Antibody (3F2), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 6 publications

Brand:  Novus Biologicals NB100-74435-0.025mg

 View more versions of this product

Product Code. 18566321

  • ONLINE EXCLUSIVE PRICE
    226.00 €
    213.15 € / 25µg
    Save 12.85 €
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Histone H2AX (p Ser139) Monoclonal antibody specifically detects Histone H2AX (p Ser139) in Human, Mouse, Bovine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Histone H2AX (p Ser139)
Monoclonal
1 mg/mL
Western Blot 1 μg/mL, Simple Western 10 μg/mL, Flow Cytometry 1 μg 106 cells, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 2 - 4 μg/mL, Immunohistochemistry-Paraffin 1:10 - 1:500
H2A.X, H2A/X, H2AFX
Mouse
15 kDa
0.025 mg
Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Mitotic Regulators, Phospho Specific
3014
Human
Purified
Western Blot
3F2
Unconjugated
P16104
H2AFX
This Histone H2AX [p Ser139] Antibody (3F2) was developed against a synthetic peptide sequence surrounding phosphorylated Ser139.
Protein G purified
RUO
Primary
In Western blot this antibody detects ∼17 kDa protein representing phosphorylated H2AX in gamma irradiated HeLa cell lysate. In immunofluorescence procedures, recognizes phosphorylated H2AX in gamma irradiated HeLa cells. ELISA of phosphorylated H2AX can also be performed. Used in IHC to successfully detect H2A.X pSer140 in postnatal mouse lung section.
Store at -20°C. Avoid freeze/thaw cycles.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only