Learn More
LAPTM4B Antibody (CL5287) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-44245
Product Code. 30286646
  • 638.00 € / 100µL
Description
LAPTM4B Monoclonal antibody specifically detects LAPTM4B in Human samples. It is validated for Immunohistochemistry (Paraffin),Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LAPTM4B
Monoclonal
LYOPH
Immunohistochemistry-Paraffin 1:200 - 1:500
LAPTM4beta, LC27, lysosomal associated protein transmembrane 4 beta, lysosomal protein transmembrane 4 beta, lysosomal-associated transmembrane protein 4B, Lysosome-associated transmembrane protein 4-beta
This antibody was generated using a synthetic peptide of Q86VI4, with the exact immunogen sequence remaining proprietary.
100 μL
Stem Cell Markers
55353
Human
Purified
Immunohistochemistry (Paraffin), Immunohistochemistry
CL5287
Unconjugated
Lyophilized in 5% Trehalose.
Mouse
Protein A purified
RUO
Primary
Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL.
Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles.
IgG2b