Learn More
Histone H3 Antibody (1B1-B2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
241.00 € - 623.00 €
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|
18795573
|
Novus Biologicals
NBP2-36468SS |
241.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18787513
|
Novus Biologicals
NBP2-36468 |
623.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Histone H3 Monoclonal specifically detects Histone H3 in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), Single Cell Western.For Research Use Only