missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3, Trimethyl Lys36 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NB21-1253C
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, C. elegans samples. It is validated for Western Blot.
Spezifikation
| Histone H3 (Trimethyl Lys36) | |
| Polyclonal | |
| Western Blot | |
| Q71DI3 | |
| H3C14 | |
| This Histone H3 [Trimethyl Lys36] antibody was raised against synthetic trimethylated peptide surrounding Lysine 36 of human Histone H3.2 [Swiss Prot Q71DI3]. | |
| 0.05 mL | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| 126961 | |
| Human, Mouse, C. elegans | |
| IgG |
| Western Blot | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology. | |
| Store at 4C in the dark. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur