missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Benzamidine FF (high sub) Columns

Product Code. 10774357 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 5mL
2 x 1mL
5 x 1mL
Flow Rate:
<20 mL/min.
<4 mL/min.
Diameter (Metric) Inner:
0.7cm
1.6cm
Unit Size:
Each
Pack of 2
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Flow Rate Diameter (Metric) Inner unitSize
10774357 2 x 1mL <4 mL/min. 0.7cm Pack of 2
11723199 1 x 5mL <20 mL/min. 1.6cm Each
10450135 5 x 1mL <4 mL/min. 0.7cm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10774357 Supplier Cytiva Supplier No. 17514302

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Remove or purify serine proteases such as thrombin. Cytiva Life Sciences™ HiTrap™ Benzamidine FF (high sub) Columns are prepacked with Benzamidine Sepharose 4 Fast Flow (high sub).

  • Prepacked with Benzamidine Sepharose™ 4 Fast Flow (high sub) media
  • Protein binding capacity ≥35mg trypsin/mL medium
  • Effective removal of thrombin and factor Xa after tag cleavage of recombinant proteins

TRUSTED_SUSTAINABILITY

Specifications

Quantity 2 x 1mL
Flow Rate <4 mL/min.
Particle Size 90 μm
For Use With (Equipment) Syringe, peristaltic pump or liquid chromato graphy system such as AKTA™ design or FPLC system
Bed Volume 1mL
Product Type Benzamidine FF Column
Diameter (Metric) Inner 0.7cm
For Use With (Application) Removal or purification of Serine Proteases such as Thrombin
Height (English) 0.98 in.
Height (Metric) 2.5cm
Ligand Type >12μM p-aminobenzamidine/mL medium
Material Polypropylene
Max. Pressure 42psi
pH Range 2 to 8
Support Type 4% Highly Cross-linked Agarose
Sample Type Serine Proteases such as Thrombin
Content And Storage 2° to 8°C
Show More Show Less
compliance-icons
Product Identifier
  • HiTrap Benzamidine FF (high sub) 1 ml, 2 x 1 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.