missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Prepacked with Benzamidine Sepharose™ 4 Fast Flow (high sub) media
- Protein binding capacity ≥35mg trypsin/mL medium
- Effective removal of thrombin and factor Xa after tag cleavage of recombinant proteins
Specifications
Specifications
| Quantity | 1 x 5mL |
| Flow Rate | <20 mL/min. |
| Particle Size | 90 μm |
| For Use With (Equipment) | Syringe, peristaltic pump or liquid chromato graphy system such as AKTA™ design or FPLC system |
| Bed Volume | 5mL |
| Product Type | Benzamidine FF Column |
| Diameter (Metric) Inner | 1.6cm |
| For Use With (Application) | Removal or purification of Serine Proteases such as Thrombin |
| Height (English) | 0.98 in. |
| Height (Metric) | 2.5cm |
| Show More |
Safety and Handling
Product Identifier
- HiTrap Benzamidine FF (high sub) 5 ml, 1 x 5 ml
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- P235-Keep cool.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- MIXTURE LIST-Contain: ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?