missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HIV1 gp120 (group M, subtype CRF07_BC) Recombinant Rabbit Monoclonal Antibody (11), HRP, Invitrogen™
GREENER_CHOICE

Product Code. p-200064018
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30019723 100 μg 100µg
30019646 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30019723 Supplier Invitrogen™ Supplier No. MA552290

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Human immunodeficiency virus type 1 (HIV-1) entry into target cells is directed by the envelope (Env) glycoproteins, which are present on the surface of HIV-1 virion or infected cells in the form of trimers consisting of gp120/gp41 complexes. The surface subunit, gp120, initiates the entry process by interacting sequentially with the CD4 receptor and a co-receptor CCR5 or CXCR4, thereby inducing a conformational change that allows the transmembrane (TM) gp41 subunit to mediate fusion between viral and target cell membranes. Cleavage of Env into its gp120 and gp41 components is necessary for activation of its fusogenic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIV1 gp120 (group M, subtype CRF07_BC)
Applications ELISA
Classification Recombinant Monoclonal
Clone 11
Concentration 100 μg/mL
Conjugate HRP
Formulation PBS with no preservative
Gene Alias HIV Glycoprotein 120; HIV-1 Glycoprotein 120; Human immunodeficiency virus
Host Species Rabbit
Immunogen Recombinant HIV-1 gp120 protein (Ala29-Glu502).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.