missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIV1 p55 Monoclonal Antibody (3A1)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA17587
This item is not returnable.
View return policy
Description
MA1-7587 detects HIV1 from virus samples. MA1-7587 has been successfully used in ELISA, Western blot, and immunoprecipitation procedures. The MA1-7587 immunogen is a purified recombinant HIV1 p55/17 protein.
HIV-1 Gag p55 is a precursor protein of several proteins that form the core structure of AIDS virus, indispensable to its reproduction. This protein is digested by HIV-1 protease, first into intermediate products p41 and p15. Then p41 is digested into matrix protein p17 and capsid protein p24. Protein p15 is further digested into nucleocapsid protein p7 and to p6 and p1, both of unknown function.
Specifications
| HIV1 p55 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.01% sodium azide; pH 7.2 | |
| Mouse | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA, Immunoprecipitation, Western Blot | |
| 3A1 | |
| Unconjugated | |
| HIV p55 | |
| Purified recombinant HIV1 p55/17 protein. | |
| 100 μg | |
| Primary | |
| 4°C short term, -80°C long term | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction