missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HJ 1052-2019 Pesticides Mixture 484 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Ametryn∣Atraton∣Atrazine∣Prometon∣Prometryn∣Propazine∣Secbumeton∣Simazine∣Simetryn∣Terbuthylazine∣Terbutryn |
| CAS | 834-12-8, 1610-17-9, 1912-24-9, 1610-18-0, 7287-19-6, 139-40-2 |
| Analyte or Component Names | Ametryn∣Atraton∣Atrazine∣Prometon∣Prometryn∣Propazine∣Secbumeton∣Simazine∣Simetryn∣Terbuthylazine∣Terbutryn |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Ametryn 100 μg/mL∣Atraton 100 μg/mL∣Atrazine 100 μg/mL∣Prometon 100 μg/mL∣Prometryn 100 μg/mL∣Propazine 100 μg/mL∣Secbumeton 100 μg/mL∣Simazine 100 μg/mL∣Simetryn 100 μg/mL∣Terbuthylazine 100 μg/mL∣Terbutryn 100 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?