missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HJ 976-2018 9 Benzenes 1000 μg/mL in Methanol, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| CAS | 71-43-2, 100-41-4, 98-82-8, 108-38-3, 95-47-6, 103-65-1 |
| Quantity | 1 mL |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction