missing translation for 'onlineSavingsMsg'
Learn More

HJC0152 hydrochloride, MedChemExpress

Product Code. 16473098
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473098 10 mg 10mg
16459656 50 mg 50mg
16454396 100 mg 100mg
16412626 25 mg 25mg
16405837 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16473098 Supplier Medchem Express Supplier No. HY100602

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HJC0152 hydrochloride is a signal transducers and activators of transcription 3 (STAT3) inhibitor.

HJC0152 hydrochloride is a signal transducers and activators of transcription 3 (STAT3) inhibitor.

Specifications

Chemical Name or Material HJC0152 hydrochloride
CAS 1420290-99-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C15H14Cl3N3O4
Quantity 10 mg
Solubility Information DMSO : 1 mg/mL (2.46 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C([N+]([O-])=O)C=C1Cl)C2=CC(Cl)=CC=C2OCCN.[H]Cl
Molecular Weight (g/mol) 406.65
Formula Weight 406.65
Percent Purity 98.95%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.