missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15656-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the ARHGAP family of Rho/Rac/Cdc42-like GTPase activating proteins. The encoded protein interacts with the Ras-related protein Gem through its N-terminal domain. Separately, it interacts with RhoA through a RhoGAP domain, and stimulates RhoA-dependent GTPase activity. Alternatively spliced variants that encode different protein isoforms have been described but their full-length nature has not been determined.Specifications
| HK1 | |
| Polyclonal | |
| Unconjugated | |
| HK1 | |
| Hk1 | |
| HK1 Fusion Protein Ag8008 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
| Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P05708, P17710, P19367 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15275, 25058, 3098 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction