missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD115 (c-fms) Monoclonal Antibody (AFS98), NovaFluor™ Yellow 730, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen™ M041T03Y07-A
This item is not returnable.
View return policy
Description
CSF1R (FMS) is involved in the production, differentiation, and function of macrophages, and is implicated in promyelocytic leukemias. Ligand binding activates the receptor kinase through a process of oligomerization and transphosphorylation. CSF1R is a tyrosine kinase transmembrane receptor and member of the CSF1/PDGF receptor family of tyrosine-protein kinases. Mutations in the gene encoding CSF1R have been associated with a predisposition to myeloid malignancy.Tyrosine-protein kinase that acts as cell-surface receptor for CSF1 and IL34 and plays an essential role in the regulation of survival, proliferation and differentiation of hematopoietic precursor cells, especially mononuclear phagocytes, such as macrophages and monocytes. Promotes the release of proinflammatory chemokines in response to IL34 and CSF1, and plays an important role in innate immunity and in inflammatory processes. The first intron of the CSF1R gene contains a transcriptionally inactive ribosomal protein L7 processed pseudogene oriented in the opposite direction. Mutations in the CSF1R gene have been associated with a predisposition to myeloid malignancy.
Specifications
| CD115 (c-fms) | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P09581 | |
| Csf1r | |
| Affinity chromatography | |
| RUO | |
| 12978 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid | |
| IgG2a κ |
| Flow Cytometry | |
| AFS98 | |
| NovaFluor Yellow 730 | |
| Csf1r | |
| AI323359; CD115; CD115 antigen; C-FMS; colony stimulating factor 1 receptor; colony stimulating factor 1 receptor, formerly McDonough feline sarcoma viral (v-fms) oncogene homolog; CSF-1 receptor; CSF1R; CSF-1R; CSF-1-R; CSFIR; Csfmr; CSFR; EC 2.7.10.1; FIM2; Fim-2; FMS; fms proto-oncogene; HDLS; kinase CSFR; macrophage colony stimulating factor I receptor; macrophage colony-stimulating factor 1 receptor; McDonough feline sarcoma viral (v-fms) oncogene homolog; M-CSFR; M-CSF-R; mrfms; OTTHUMP00000224093; protein-tyrosine kinase; proto-oncogene c-Fms; proto-oncogene fms | |
| Rat | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction