missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-A2 Monoclonal Antibody (BB7.2), Super Bright™ 436, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
159.00 € - 362.00 €
Specifications
| Antigen | HLA-A2 |
|---|---|
| Clone | BB7.2 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16391324
|
eBioscience
62-9876-41 |
25 Tests | |||||||
16301334
![]() |
Invitrogen™
62-9876-42 |
100 Tests |
362.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This BB7.2 monoclonal antibody recognizes the human major histocompatibility complex (MHC) class I molecule HLA-A2, which is a member of the HLA-A family. MHC class I exists as a single alpha chain composed of three domains. This molecule associates with
HLA-A belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. Class I molecules play a central role in the immune system by presenting peptides derived from the endoplasmic reticulum lumen. They are expressed in nearly all cells. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon 1 encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domains, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exons 6 and 7 encode the cytoplasmic tail. Polymorphisms within exon 2 and exon 3 are responsible for the peptide binding specificity of each class one molecule. Typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. Hundreds of HLA-A alleles have been described. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.Specifications
| HLA-A2 | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P04439 | |
| HLA-A | |
| Primary | |
| Antibody |
| BB7.2 | |
| Flow Cytometry | |
| Super Bright 436 | |
| Mouse | |
| Human | |
| 3105 | |
| IgG2b κ | |
| 4° C, store in dark, DO NOT FREEZE! | |
| HLA-A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title