missing translation for 'onlineSavingsMsg'
Learn More

HLA-A2 Mouse anti-Human, FITC, Clone: BB7.2, BD

Mouse Monoclonal Antibody

110.21 € - 257.77 €

Spécification

Antigen HLA-A2
Clone BB7.2
Concentration 0.5mg/mL
Applications Flow Cytometry
Classification Monoclonal
Consulter d'autres spécifications

Produits 2
Code produit Marque Quantity Prix Quantité et disponibilité  
Code produit Marque Quantity Prix Quantité et disponibilité  
15840779
missing translation for 'viewDocuments'
BD Biosciences
561107
25 μg
ONLINE EXCLUSIVE PRICE
115.00 €
110.21 € / Each
Save 4.79 €
4% Off
Voir les produits de remplacement
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
15836598
missing translation for 'viewDocuments'
BD Biosciences
551285
0.1 mg
ONLINE EXCLUSIVE PRICE
268.00 €
257.77 € / Each
Save 10.23 €
3% Off
Voir les produits de remplacement
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The monoclonal antibody BB7.2 specifically binds to the α subunit of the human leukocyte antigen-A2 (HLA-A2), a class I molecule of the major histocompatibility complex (MHC). The MHC gene locus encodes a group of highly polymorphic, cell-surface proteins that play a broad role in the immune response to protein antigens. MHC molecules function by binding and presenting small antigenic protein fragments to antigen-specific receptors expressed by T cells (TCR). Human (human leukocyte antigen/HLA) MHC molecules comprise two major classes, MHC class I and class II. Functionally, class I MHC molecules can bind peptides derived from intracellular antigens (eg, viral and some bacterial antigens) that are specifically recognized by CD8+ T cells, and class II MHC molecules recognize antigens derived from pathogens multiplying in intracellular vesicles, and those derived from ingested extracellular bacteria. When presented on the cell surface by the MHC class II molecules, these antigens are recognized by CD4+ T cells. TCR recognize both processed peptides bound to MHC, as well as regions of the MHC molecule itself. CD4 and CD8 accessory molecules strengthen formation of the TCR-MHC complex through their interaction with non-polymorphic regions of the MHC molecule.

Spécification

Spécification

HLA-A2
0.5mg/mL
Monoclonal
Mouse
Human
IgG2b κ
Affinity Purified
BB7.2
Flow Cytometry
FITC
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Vidéos
FDS
Documents

Documents

Certificats

For Research Use Only.

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis