Learn More
HLA-B7 Monoclonal Antibody (BB7.1), PE, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
139.00 € - 371.00 €
Specifications
| Antigen | HLA-B7 |
|---|---|
| Clone | BB7.1 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16340624
|
Affymetrix eBioscience
12-5917-41 |
25 Tests | |||||||
16350624
![]() |
Invitrogen™
12-5917-42 |
100 Tests |
371.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: This BB7.1 monoclonal antibody recognizes the human major histocompatibility complex MHC class I molecule HLA-B7, which is a member of the HLA-B family. MHC class I exists as a heterodimer composed of a specific alpha chain associated with a beta 2-microglobulin. MHC class I molecules are expressed on all nucleated cells. Applications Reported: This BB7.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This BB7.1 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser
HLA-B7 is one of the polymorphic proteins of human MHC Class I found on all nucleated cells. This surface protein is a heterodimer consisting of the specific transmembrane HLA-B chain paired with Beta-2 microglobulin. Classical MHC Class I molecules play a central role in adaptive immunity by presenting to T cells antigenic peptides derived either from the cell itself (classical presentation) or extracellular antigens (cross-presentation). The ligands of MHC class I include T cell receptor (TCR) and a costimulatory molecule CD8. Hundreds of HLA-B alleles have been described. Typing for these polymorphisms is routinely done for transplantation. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.Specifications
| HLA-B7 | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P01889 | |
| HLA-B | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| BB7.1 | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| Human | |
| 3106 | |
| Papain solubilized HLA-B7 | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! | |
| HLA-B |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.