missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HLA-B7 Recombinant Mouse Monoclonal Antibody (BB7.1)

Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556743
This item is not returnable.
View return policy
Description
HLA-B7 Recombinant Monoclonal Antibody for Flow
HLA-B7 is one of the polymorphic proteins of human MHC Class I found on all nucleated cells. This surface protein is a heterodimer consisting of the specific transmembrane HLA-B chain paired with Beta-2 microglobulin. Classical MHC Class I molecules play a central role in adaptive immunity by presenting to T cells antigenic peptides derived either from the cell itself (classical presentation) or extracellular antigens (cross-presentation). The ligands of MHC class I include T cell receptor (TCR) and a costimulatory molecule CD8. Hundreds of HLA-B alleles have been described. Typing for these polymorphisms is routinely done for transplantation. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifications
| HLA-B7 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P01889 | |
| HLA-B | |
| The immunogen used to generate this antibody corresponds to human HLA-B7. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| BB7.1 | |
| Unconjugated | |
| HLA-B | |
| A29 protein; antigen presenting molecule; AS; Aw-33; Aw-74; B-4901; B-5001; Bw-47; Bw-50; CDABP0067; class I MHC, HLA-B antigen; DAQB-90C11.16; epididymis secretory protein Li 83; FLJ26655; HEL-S-83; histocompatibility antigen; HLA A; HLA -B; HLA B-40011; HLA B44; hla b7; HLA class I antigen; HLA class I antigen HLA-B; HLA class I histocompatibility antigen, A alpha chain; HLA class I histocompatibility antigen, A-1 alpha chain; HLA class I histocompatibility antigen, A-30 alpha chain; HLA class I histocompatibility antigen, A-33 alpha chain; HLA class I histocompatibility antigen, A-74 alpha chain; HLA class I histocompatibility antigen, B alpha chain; HLA class I histocompatibility antigen, B-47 alpha chain; HLA class I histocompatibility antigen, B-50 alpha chain; HLA class I histocompatibility antigen, B-82 alpha chain; HLA locus; HLA molecule; HLAA; HLA-A; HLA-A locus alpha 2 domain; HLA-A null; HLA-A11; HLA-A33; HLA-Aw33.1; HLAB; HLA-B; HLA-B 1502; HLA-B alpha chain; HLA-B alpha chain (B*2706); HLA-B alpha chain (B*5002); HLA-B alpha-chain; HLA-B*45ZJ; HLA-B15; HLA-B35; HLA-B35 antigen; HLA-B-3506; HLA-B39; HLA-B-3905; HLA-B49; HLA-B50; HLA-B55; HLA-B-5502; HLA-B-5602; HLA-B59; HLA-B61; HLA-Bw50 antigen; HLA-Cw; HLA-DQB1; HLA-DRB1; Human leukocyte antigen A; hypothetical protein; lecocyte antigen B; Leukocyte Antigen; leukocyte antigen class I-A; leukocyte antigen class I-B; Leukoocyte Antigen; lymphocyte antigen; major histocompatability complex B antigen; major histocompatibility complex, class I, A; major histocompatibility complex, class I, B; MCH class I antigen; MHC 1; MHC antigen; MHC class 1 antigen; MHC class I alpha chain; MHC class I anitgen; MHC class I anti; MHC Class I antigen; MHC class I antigen A*30; MHC class I antigen A*33; MHC class I antigen A*74; MHC class I antigen B*47; MHC class I antigen B*50; MHC class I antigen B*82; MHC class I antigen GN00104; MHC class I antigen heavy chain; MHC class I antigen HLA-A heavy chain; MHC class I antigen HLA-A33; MHC class I antigen HLA-B alpha chain; MHC class I antigen HLA-B heavy chain; MHC class I antigen null protein; MHC class I antigen SHCHA; MHC class I antigene; MHC class I human leukocyte antigen; MHC class I molecule; MHC class I protein; MHC class II antigen; MHC class l antigen; MHC classI antigen; MHC HLA-B cell surface glycoprotein; MHC HLA-B transmembrane glycoprotein; MHC HLA-B27d; MHC HLA-B27-HS; MHC HLA-B39N; MHC1; SPDA1; truncated class I antigen; truncated MHC class I antigen | |
| Mouse | |
| Protein A | |
| RUO | |
| 3106 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction