missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DMB Antibody (SR1484), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00 € - 449.00 €
Specifications
| Antigen | HLA DMB |
|---|---|
| Clone | SR1484 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50 |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18691314
|
Novus Biologicals
NBP3-22457-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18692025
|
Novus Biologicals
NBP3-22457-50ul |
50 μg |
338.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLA DMB Monoclonal antibody specifically detects HLA DMB in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
| HLA DMB | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| class II histocompatibility antigen, M beta chain, D6S221E, DMB, major histocompatibility complex, class II, DM beta, MHC class II antigen DMB, MHC class II antigen HLA-DM beta chain, MHC class II HLA-DMB, Really interesting new gene 7 protein, RING7HLA class II histocompatibility antigen, DM beta chain | |
| A synthesized peptide derived from human HLA DMB (Uniprot #: P28068) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1484 | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Asthma, Diabetes Research, Immunology | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 3109 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title