missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DQ/DR/DP Antibody (rHLA-Pan/8847), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23657-20ug
This item is not returnable.
View return policy
Description
HLA DQ/DR/DP Monoclonal antibody specifically detects HLA DQ/DR/DP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| HLA DQ/DR/DP | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha | |
| Priess human B cell line | |
| 20 μg | |
| Adaptive Immunity, Asthma, Diabetes Research, Immunology | |
| 3117 | |
| Store at 4°C. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rHLA-Pan/8847 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction