Learn More
HLA-DRA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17221-1-AP-20UL
 View more versions of this product
Product Code. 16847074
  • 145.00 € / 20µL
Description
Expression of the invariant chain of the HLA-DR antigen (CD74) by B-lymphocytes begins in the pre-B stage but lost before the final differentiation to plasma cells. This protein is expressed in B-lymphocytes and many B-cell lymphomas.
Specifications
Specifications
HLA-DRA
Polyclonal
Unconjugated
HLA-DRA
HLA DRA, HLA DRA1, HLA-DRA, MHC class II antigen DRA
Rabbit
Antigen Affinity Chromatography
RUO
3122
-20°C
Liquid
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P01903
HLA-DRA
HLA-DRA Fusion Protein Ag11044
20 μL
Primary
Human, Rat, Mouse