missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HLA-F Recombinant Rabbit Monoclonal Antibody (24GB3830)

Rabbit Recombinant Monoclonal Antibody
146.00 € - 500.00 €
Specifications
| Antigen | HLA-F |
|---|---|
| Clone | 24GB3830 |
| Concentration | 0.387 mg/mL |
| Content And Storage | -20°C |
| Applications | Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276149
![]() |
Invitrogen™
MA557037 |
100 μL |
500.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275647
|
Invitrogen™
MA557038 |
20 μL |
146.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLA-F Recombinant Monoclonal Antibody for Western Blot, ICC/IF
HLA-F belongs to the HLA class I heavy chain paralogues. It is a non-classical heavy chain that forms a heterodimer with a beta-2 microglobulin light chain, with the heavy chain anchored in the membrane. Unlike most other HLA heavy chains, this molecule is localized in the endoplasmic reticulum and Golgi apparatus, with a small amount present at the cell surface in some cell types. It contains a divergent peptide-binding groove, and is thought to bind a restricted subset of peptides for immune presentation.HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.Specifications
| HLA-F | |
| 0.387 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P30511 | |
| 3134 | |
| A synthesized peptide derived from human HLA F (aa 25-76). | |
| Antibody |
| 24GB3830 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| HLA-F | |
| CDA12; HLA class I histocompatibility antigen, alpha chain F; HLA class I molecule; HLA F antigen; HLA-5.4; HLA-CDA12; HLAF; HLA-F; Leukocyte antigen F; major histocompatibility complex, class I, F; MHC class I antigen F; MHC class Ib antigen | |
| HLA-F | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title