Learn More
HMG-CoA Reductase/HMGCR Antibody (CL0260) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61617MFV450
Product Code. 30302345
  • 759.00 € / 0.10mL
Description
HMG-CoA Reductase/HMGCR Monoclonal antibody specifically detects HMG-CoA Reductase/HMGCR in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HMG-CoA Reductase/HMGCR
Monoclonal
mFluor Violet 450 SE
3-hydroxy-3-methylglutaryl CoA reductase (NADPH), 3-hydroxy-3-methylglutaryl-CoA reductase, 3-hydroxy-3-methylglutaryl-Coenzyme A reductase, EC 1.1.1, EC 1.1.1.34, HMG-CoA reductase, hydroxymethylglutaryl-CoA reductase, LDLCQ3
This antibody was developed using a recombinant protein derived from P04035, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction
3156
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown
CL0260
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified