Learn More
HMG20A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12085-2-AP-150UL
 View more versions of this product
Product Code. 16889583
  • 410.00 € / 150µL
Description
Plays a role in neuronal differentiation as chromatin-associated protein. Acts as inhibitor of HMG20B. Overcomes the repressive effects of the neuronal silencer REST and induces the activation of neuronal-specific genes. Involved in the recruitment of the histone methyltransferase KMT2A/MLL1 and consequent increased methylation of histone H3 lysine 4.
Specifications
Specifications
HMG20A
Polyclonal
Unconjugated
HMG20A
high mobility group 20A, HMG box containing protein 20A, HMG20A, HMGX1, HMGXB1
Rabbit
Antigen Affinity Chromatography
RUO
10363, 315689, 66867
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9DC33, Q9NP66
Hmg20a
HMG20A Fusion Protein Ag2718
150 μL
Primary
Human, Mouse, Rat