Learn More
HMGA2 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-20795-100UL
Description
The HMGA2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human HMGA2. This antibody recognizes human, mouse, rat antigen. The HMGA2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| HMGA2 | |
| 1000 μg/mL | |
| BABL, high mobility group AT hook 2, HMGA2, HMGI C, HMGIC, LIPO, STQTL9 | |
| 100 μL | |
| 8091 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |