missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGCS2 Antibody (SR1885), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22333-50ul
This item is not returnable.
View return policy
Description
HMGCS2 Monoclonal antibody specifically detects HMGCS2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| HMGCS2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1885 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| 3-hydroxy-3-methylglutaryl coenzyme A synthase, 3-hydroxy-3-methylglutaryl-CoA synthase 2 (mitochondrial), 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 2 (mitochondrial), EC 2.3.3.10, HMG-CoA synthase, hydroxymethylglutaryl-CoA synthase, mitochondrial, mitochondrial 3-hydroxy-3-methylglutaryl-CoA synthase | |
| A synthesized peptide derived from human HMGCS2 (Uniprot #: P54868) | |
| 50 μg | |
| Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 3158 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction