missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGN3/TRIP7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 572.00 €
Specifications
| Antigen | HMGN3/TRIP7 |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18474311
|
Novus Biologicals
NBP1-86904-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18259717
|
Novus Biologicals
NBP1-86904 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HMGN3/TRIP7 Polyclonal specifically detects HMGN3/TRIP7 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| HMGN3/TRIP7 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| DKFZp686E20226, high mobility group nucleosomal binding domain 3, high mobility group nucleosome-binding domain-containing protein 3, high-mobility group protein HMGN3, PNAS-25, thyroid hormone receptor interacting protein 7, thyroid hormone receptor interactor 7, Thyroid receptor-interacting protein 7, TR-interacting protein 7, TRIP-7, TRIP7FLJ42187 | |
| HMGN3 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 9324 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KPEPKPRKTSAKKEPGAKISRGAKGKKEEKQEAGKEGTAPSENGETKAEEAQKTESVDNEGE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title