missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF-4 alpha/NR2A1 Antibody (SR1937), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22529-50ul
This item is not returnable.
View return policy
Description
HNF-4 alpha/NR2A1 Monoclonal antibody specifically detects HNF-4 alpha/NR2A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
Specifications
| HNF-4 alpha/NR2A1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| SR1937 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
| FLJ39654, hepatic nuclear factor 4 alpha, hepatocyte nuclear factor 4, alpha, hepatocyte nuclear factor 4-alpha, HNF4a7, HNF-4-alpha, HNF4alpha10/11/12, HNF4HNF4a8, MODY, MODY1, NR2A1HNF4a9, NR2A21, Nuclear receptor subfamily 2 group A member 1, TCF, TCF-14, TCF14HNF4alpha, Transcription factor 14, Transcription factor HNF-4, transcription factor-14 | |
| A synthesized peptide derived from human HNF-4 alpha/NR2A1 (Uniprot #: P41235) | |
| 50 μg | |
| Phospho Specific, Stem Cell Markers | |
| 3172 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction