missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97829-100ul
This item is not returnable.
View return policy
Description
hnRNP A1 Polyclonal antibody specifically detects hnRNP A1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| hnRNP A1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| Helix-destabilizing protein, heterogeneous nuclear ribonucleoprotein A1, heterogeneous nuclear ribonucleoprotein A1B protein, heterogeneous nuclear ribonucleoprotein B2 protein, heterogeneous nuclear ribonucleoprotein core protein A1, hnRNP A1, hnRNP core protein A1, hnRNPA1, hnRNP-A1, HNRPA1MGC102835, nuclear ribonucleoprotein particle A1 protein, single-strand DNA-binding protein UP1, Single-strand RNA-binding protein | |
| Produced in rabbits immunized with E. coli-derived Human hnRNP A1 fragment. | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 3178 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur