missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP K Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-24531
This item is not returnable.
View return policy
Description
hnRNP K Polyclonal specifically detects hnRNP K in Human, Mouse, Rat, Opossum, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| hnRNP K | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| HNRNPK | |
| A portion of amino acid 1 to 50 of human hnRNPK was used as immunogen for the antibody. | |
| 0.1 mg | |
| Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 3190 | |
| Human, Mouse, Rat, Opossum, Primate | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 2-5 ug/ml. Use reported by customer review, Immunohistochemistry, Immunohistochemistry-Paraffin 5-10 ug/ml | |
| dC-stretch binding protein, FLJ41122, heterogeneous nuclear ribonucleoprotein K, hnRNP K, transformation upregulated nuclear protein, Transformation up-regulated nuclear protein, TUNPHNRPKCSBP | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen for the Importin-8 antibody is 100% homologous in human and chimpanzee and monkey, 97% incow and dog, 92% in mouse, and 86% homologous inchicken and opossum. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction