Learn More
hnRNP L Antibody (4D11), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB120-6106JF646
Product Code. 18746801
  • 568.00 € / 0.10mL
Description
hnRNP L Monoclonal antibody specifically detects hnRNP L in Human, Mouse, Rat, Amphibian, Bovine, Chicken, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
hnRNP L
Monoclonal
Janelia Fluor 646
FLJ35509, heterogeneous nuclear ribonucleoprotein L, hnRNP L, hnRNP-L, HNRPL
Human hnRNP proteins from HeLa cells purified by affinity chromatography on ssDNA agarose.
0.1 mL
Cancer, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers
3191
Store at 4°C in the dark.
IgG1
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
4D11
50mM Sodium Borate
Mouse
Protein G purified
RUO
Primary
Human, Mouse, Rat, Amphibian, Bovine
Purified