Learn More
Invitrogen™ hnRNP L Monoclonal Antibody (4D11)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA546550
Product Code. 17810316
  • 433.00 € / 50µg
Description
Perform heat mediated antigen retrieval with citrate buffer pH 6 before commencing with IHC staining protocol. Western blot detects a band of approximately 64 kDa (predicted molecular weight: 64 kDa).
Heterogeneous nuclear RNAs (hnRNAs) which include mRNA precursors and mature mRNAs are associated with specific proteins to form heterogenous ribonucleoprotein (hnRNP) complexes. Heterogeneous nuclear ribonucleoprotein L is among the proteins that are stably associated with hnRNP complexes and along with other hnRNP proteins is likely to play a major role in the formation, packaging, processing, and function of mRNA. Heterogeneous nuclear ribonucleoprotein L is present in the nucleoplasm as part of the HNRP complex. HNRP proteins have also been identified outside of the nucleoplasm. Exchange of hnRNP for mRNA-binding proteins accompanies transport of mRNA from the nucleus to the cytoplasm. Since HNRP proteins have been shown to shuttle between the nucleus and the cytoplasm, it is possible that they also have cytoplasmic functions. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
hnRNP L
Monoclonal
1 mg/mL
PBS with no preservative
F1LQ48, P14866, Q8R081
HNRNPL
Human hnRNP proteins, from HeLa cells, purified by affinity chromatography on ssDNA agarose.
50 μg
Primary
Human, Mouse, Rat, Amphibian
Antibody
IgG1
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
4D11
Unconjugated
HNRNPL
C79783; D830027H13Rik; Fblim1; FLJ35509; Heterogeneous nuclear ribonucleoprotein L; heterogenous nuclear ribonucleoprotein L; hnRNP L; Hnrnpl; hnRNP-L; Hnrpl; P/OKcl.14
Mouse
Protein A
RUO
15388, 3191, 80846
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.