missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNRPDL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 624.00 €
Specifications
| Antigen | HNRPDL |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18619025
|
Novus Biologicals
NBP2-38806-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18129767
|
Novus Biologicals
NBP2-38806 |
0.1 mL |
624.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HNRPDL Polyclonal specifically detects HNRPDL in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| HNRPDL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| O14979 | |
| 9987 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: DSSVTMEDMNEYSNIEEFAEGSKINASKNQQDDGKM | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| A+U-rich element RNA binding factor, AU-rich element RNA-binding factor, heterogeneous nuclear ribonucleoprotein D-like, HNRNP, hnRNP DL, hnRNP D-like, JKTBP2, JKTBPJKT41-binding protein, laAUF1, Protein laAUF1 | |
| HNRPDL | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title