Learn More
HO-1/HMOX1 Rabbit anti-Human, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL594-10701-100UL
Description
The HO-1/HMOX1 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human HO-1/HMOX1. This antibody recognizes human, mouse, rat antigen. The HO-1/HMOX1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| HO-1/HMOX1 | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat |
| Immunofluorescence | |
| CoraLite 594 | |
| 100 μL | |
| 3162 | |
| IgG |