missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) sulfate hydrate, 99.99%, Thermo Scientific™

Product Code. 11458966
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11458966

Brand: Thermo Scientific Alfa Aesar A17092.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15622-40-9
Molecular Formula Ho2O12S3
Molecular Weight (g/mol) 618.03
MDL Number MFCD00149740
InChI Key MKPJADFELTTXAV-UHFFFAOYSA-H
IUPAC Name diholmium(3+) trisulfate
SMILES [Ho+3].[Ho+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Quantity 5 g
Linear Formula Ho2(SO4)3·xH2O
Formula Weight 618.03 (Anhydrous)
Percent Purity 99.99%
Sensitivity Hygroscopic
Chemical Name or Material Holmium(III) sulfate hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.