missing translation for 'onlineSavingsMsg'
Learn More

Holmium Standard: Ho @ 1000 μg/mL in 5% HNO3, VHG™

Product Code. 17978449
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17978449 50 mL 50mL
17917219 250 mL 250mL
17958389 500 mL 500mL
17907219 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 17978449 Supplier VHG Labs Supplier No. VHGPHON50
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Holmium Standard: Ho @ 1000 μg/mL in 5% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Holmium
CAS 7440-60-0
Methods ICP/ICP-MS
Analyte or Component Names Holmium
Shipping Condition Room Temperature
Molecular Formula Ho
Certifications/Compliance ISO 17034
Synonym Ho
InChI Formula InChI=1S/Ho
SMILES [Ho]
IUPAC Name holmium
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 164.93
Quantity 50 mL
Formula Weight 164.93 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.