missing translation for 'onlineSavingsMsg'
Learn More

HOMEZ Rabbit anti-Human, Rat, Polyclonal, Proteintech

Product Code. 16802565 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16802565 20 μL 20µL
16892555 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16802565 Supplier Proteintech Supplier No. 239651AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

May function as a transcriptional regulator.

Specifications

Antigen HOMEZ
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HOMEZ
Gene Accession No. Q8IX15, Q8K3E9
Gene Alias HOMEZ, KIAA1443
Gene Symbols Homez
Host Species Rabbit
Immunogen HOMEZ Fusion Protein Ag21090
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 260325, 57594
Target Species Rat, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.