missing translation for 'onlineSavingsMsg'
Learn More

Homogentisic Acid, TRC

Product Code. 30008980
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008980 100 mg 100mg
30005222 250 mg 250mg
30008449 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008980 Supplier Toronto Research Chemicals Supplier No. TRCH593400100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 451-13-8
Molecular Formula C8 H8 O4
Molecular Weight (g/mol) 168.15
Synonym 2,5-Dihydroxybenzeneacetic Acid, 2,5-Dihydroxy-α-toluic Acid, 2,5-Dihydroxyphenylacetic Acid, Alcapton, Homogentisinic Acid, NSC 88940
IUPAC Name 2-(2,5-dihydroxyphenyl)acetic acid
SMILES OC(=O)Cc1cc(O)ccc1O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H8O4/c9-6-1-2-7(10)5(3-6)4-8(11)12/h1-3,9-10H,4H2,(H,11,12)
Formula Weight 168.0423
Chemical Name or Material Homogentisic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.