missing translation for 'onlineSavingsMsg'
Learn More

Homovanillic acid, MedChemExpress

Product Code. 16499066
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499066 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16499066 Supplier Medchem Express Supplier No. HYN0384
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Homovanillic acid is a dopamine metabolite found to be associated with aromatic L-amino acid decarboxylase deficiency, celiac disease, growth hormone deficiency, and sepiapterin reductase deficiency.

Homovanillic acid is a dopamine metabolite found to be associated with aromatic L-amino acid decarboxylase deficiency, celiac disease, growth hormone deficiency, and sepiapterin reductase deficiency.

Specifications

Chemical Name or Material Homovanillic acid
CAS 306-08-1
Color Gray
Purity Grade Notes Research
Molecular Formula C9H10O4
Quantity 100 mg
Synonym Vanilacetic acid
Solubility Information DMSO : ≥ 100 mg/mL (548.94 mM) ∣H2O : 8.33 mg/mL (45.73 mM; Need ultrasonic)
SMILES O=C(O)CC1=CC=C(O)C(OC)=C1
Molecular Weight (g/mol) 182.17
Formula Weight 182.17
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.